1-(3-Bromopyridin-2-yl)-2,2-dichloroethanol ≥95%

Supplier: AOBChem USA

Warning

78953-1G 78953-500MG 78953-5G 78953-25G
77473-224EA 415.09 USD
77473-224 77485-842 77496-438 77505-508
1-(3-Bromopyridin-2-yl)-2,2-dichloroethanol ≥95%
1-(3-Bromopyridin-2-yl)-2,2-dichloroethanol

Precaución: For research use only

Formula: C₇H₆BrCl₂NO
MW: 270.939 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767695
CAS Number: 2901099-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR