1-(2-Fluoro-3,4-dimethoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

78719-1G 78719-500MG 78719-5G
77473-220EA 454.23 USD
77473-220 77485-838 77496-434
1-(2-Fluoro-3,4-dimethoxyphenyl)ethanone ≥97%
1-(2-Fluoro-3,4-dimethoxyphenyl)ethanone

Precaución: For research use only

Formula: C₁₀H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD20486611
CAS Number: 158641-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR