Methyl 4-bromo-3-fluoro-5-methoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

78625-1G 78625-500MG 78625-5G 78625-25G
77473-216EA 284.28 USD
77473-216 77485-834 77496-430 77505-504
Methyl 4-bromo-3-fluoro-5-methoxybenzoate ≥95%
Methyl 4-bromo-3-fluoro-5-methoxybenzoate

Precaución: For research use only

Formula: C₉H₈BrFO₃
MW: 263.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23707521
CAS Number: 1427437-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR