2-(3,6-Difluoro-2-(methylthio)phenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

78567-1G 78567-500MG 78567-5G
77473-212EA 690.1 USD
77473-212 77485-830 77496-426
2-(3,6-Difluoro-2-(methylthio)phenyl)-1,3-dioxolane ≥97%
2-(3,6-Difluoro-2-(methylthio)phenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₀F₂O₂S
MW: 232.247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34816222
CAS Number: 2918959-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR