4-Bromo-5-iodo-1-methyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

78190-1G 78190-500MG 78190-5G
77473-204EA 701.43 USD
77473-204 77485-822 77496-416
4-Bromo-5-iodo-1-methyl-1H-pyrazole ≥97%
4-Bromo-5-iodo-1-methyl-1H-pyrazole

Precaución: For research use only

Formula: C₄H₄BrIN₂
Storage Temperature: Ambient
MDL Number: MFCD30615838
CAS Number: 2138196-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR