2-Fluoro-3-iodo-5-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

78090-1G 78090-500MG 78090-5G 78090-25G
77473-200EA 415.09 USD
77473-200 77485-818 77496-410 77505-494
2-Fluoro-3-iodo-5-methoxybenzaldehyde ≥97%
2-Fluoro-3-iodo-5-methoxybenzaldehyde

Precaución: For research use only

Formula: C₈H₆FIO₂
Storage Temperature: Ambient
MDL Number: MFCD34767329
CAS Number: 2383184-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR