1-(2-Fluoro-5-methoxyphenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

78046-1G 78046-500MG 78046-5G
77473-196EA 541.78 USD
77473-196 77485-814 77496-406
1-(2-Fluoro-5-methoxyphenyl)ethanol ≥97%
1-(2-Fluoro-5-methoxyphenyl)ethanol

Precaución: For research use only

Formula: C₉H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD26136304
CAS Number: 1502127-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR