1,5-Difluoro-2-methyl-3-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

77971-1G 77971-500MG 77971-5G 77971-25G
77473-192EA 241.02 USD
77473-192 77485-810 77496-402 77505-488
1,5-Difluoro-2-methyl-3-nitrobenzene ≥97%
1,5-Difluoro-2-methyl-3-nitrobenzene

Precaución: For research use only

Formula: C₇H₅F₂NO₂
MW: 173.1169 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22557859
CAS Number: 1188412-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR