5-Bromo-1-ethyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

77969-1G 77969-500MG 77969-5G 77969-25G
77473-188EA 238.96 USD
77473-188 77485-806 77496-398 77505-484
5-Bromo-1-ethyl-1H-pyrazole ≥97%
5-Bromo-1-ethyl-1H-pyrazole

Precaución: For research use only

Formula: C₅H₇BrN₂
MW: 175.026 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18447684
CAS Number: 1268334-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR