6-Fluoro-2-iodo-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

77884-1G 77884-500MG 77884-5G
77473-184EA 508.82 USD
77473-184 77485-802 77496-394
6-Fluoro-2-iodo-3-methoxybenzaldehyde ≥97%
6-Fluoro-2-iodo-3-methoxybenzaldehyde

Precaución: For research use only

Formula: C₈H₆FIO₂
MW: 280.035 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767253
CAS Number: 2090466-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR