4-Bromo-5-(methylthio)thiophene-2-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

77857-1G 77857-500MG 77857-5G 77857-25G
77473-180EA 415.09 USD
77473-180 77485-798 77496-390 77505-478
4-Bromo-5-(methylthio)thiophene-2-carbaldehyde ≥97%
4-Bromo-5-(methylthio)thiophene-2-carbaldehyde

Precaución: For research use only

Formula: C₆H₅BrOS₂
MW: 237.137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767239
CAS Number: 1160308-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR