2-(Methylthio)benzo[b]thiophene ≥97%

Supplier: AOBChem USA

Warning

77822-1G 77822-500MG 77822-5G 77822-25G
77473-172EA 415.09 USD
77473-172 77485-790 77496-382 77505-472
2-(Methylthio)benzo[b]thiophene ≥97%
2-(Methylthio)benzo[b]thiophene

Precaución: For research use only

Formula: C₉H₈S₂
MW: 180.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28060427
CAS Number: 30489-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR