2,4-Diethoxy-3-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

77243-1G 77243-500MG 77243-5G
77473-164EA 835.33 USD
77473-164 77485-782 77496-372
2,4-Diethoxy-3-fluorobenzaldehyde ≥97%
2,4-Diethoxy-3-fluorobenzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₃FO₃
MW: 212.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766934
CAS Number: 2901086-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR