4-Fluoro-2,5-dimethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

77138-1G 77138-500MG 77138-5G 77138-25G
77473-160EA 172.01 USD
77473-160 77485-778 77496-368 77505-460
4-Fluoro-2,5-dimethoxybenzaldehyde ≥97%
4-Fluoro-2,5-dimethoxybenzaldehyde

Precaución: For research use only

Formula: C₉H₉FO₃
MW: 184.1643 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19301638
CAS Number: 82830-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR