3-(Methylthio)-5-propoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

77106-1G 77106-500MG 77106-5G 77106-25G
77473-156EA 415.09 USD
77473-156 77485-774 77496-364 77505-456
3-(Methylthio)-5-propoxybenzaldehyde ≥95%
3-(Methylthio)-5-propoxybenzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₄O₂S
MW: 210.2926 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766851
CAS Number: 2918853-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR