(3-Fluoro-4-methoxy-2-(methoxymethoxy)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

76860-1G 76860-500MG 76860-5G 76860-25G
77473-152EA 415.09 USD
77473-152 77485-770 77496-358 77505-450
(3-Fluoro-4-methoxy-2-(methoxymethoxy)phenyl)(methyl)sulfane ≥97%
(3-Fluoro-4-methoxy-2-(methoxymethoxy)phenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₀H₁₃FO₃S
MW: 232.272 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766753
CAS Number: 2901066-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR