3-Fluoro-2-isopropoxy-4-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

76838-1G 76838-500MG 76838-5G 76838-25G
77473-148EA 415.09 USD
77473-148 77485-766 77496-354 77505-446
3-Fluoro-2-isopropoxy-4-methoxybenzaldehyde ≥97%
3-Fluoro-2-isopropoxy-4-methoxybenzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₃FO₃
MW: 212.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766735
CAS Number: 2901096-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR