Ethyl-2-(3-bromopyridin-4-yl)acetate ≥97%

Supplier: AOBChem USA

Warning

76719-1G 76719-500MG 76719-5G 76719-25G
77473-144EA 272.95 USD
77473-144 77485-762 77496-350 77505-442
Ethyl-2-(3-bromopyridin-4-yl)acetate ≥97%
Ethyl-2-(3-bromopyridin-4-yl)acetate

Precaución: For research use only

Formula: C₉H₁₀BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD17011773
CAS Number: 51054-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR