(2,6-Difluoro-3-methylphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

76398-1G 76398-500MG 76398-5G 76398-25G
77473-140EA 105.06 USD
77473-140 77485-758 77496-346 77505-438
(2,6-Difluoro-3-methylphenyl)trimethylsilane ≥95%
(2,6-Difluoro-3-methylphenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₀H₁₄F₂Si
MW: 200.3004 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29960055
CAS Number: 348080-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR