5-Bromo-3-chloro-2-(methoxymethoxy)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

76298-1G 76298-500MG 76298-5G 76298-25G
77473-136EA 162.74 USD
77473-136 77485-754 77496-342 77505-434
5-Bromo-3-chloro-2-(methoxymethoxy)-1,1'-biphenyl ≥95%
5-Bromo-3-chloro-2-(methoxymethoxy)-1,1'-biphenyl

Precaución: For research use only

Formula: C₁₄H₁₂BrClO₂
MW: 327.601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766421
CAS Number: 2901107-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR