4-Fluoro-5-isopropoxy-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

76075-1G 76075-500MG 76075-5G 76075-25G
77473-128EA 415.09 USD
77473-128 77485-746 77496-334 77505-428
4-Fluoro-5-isopropoxy-2-methylbenzaldehyde ≥97%
4-Fluoro-5-isopropoxy-2-methylbenzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD34766248
CAS Number: 2901086-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR