4-Fluoro-3-(methoxymethoxy)-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

75901-1G 75901-500MG 75901-5G
77473-124EA 554.14 USD
77473-124 77485-742 77496-330
4-Fluoro-3-(methoxymethoxy)-5-(trifluoromethyl)benzoic acid ≥97%
4-Fluoro-3-(methoxymethoxy)-5-(trifluoromethyl)benzoic acid

Precaución: For research use only

Formula: C₁₀H₈F₄O₄
MW: 268.1617 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766209
CAS Number: 2766311-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR