tert-Butyl 2-chloro-6-fluorobenzoate ≥95%

Supplier: AOBChem USA

Warning

75884-1G 75884-500MG 75884-5G 75884-25G
77473-120EA 146.26 USD
77473-120 77485-738 77496-326 77505-422
tert-Butyl 2-chloro-6-fluorobenzoate ≥95%
tert-Butyl 2-chloro-6-fluorobenzoate

Precaución: For research use only

Formula: C₁₁H₁₂ClFO₂
MW: 230.6631 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19543474
CAS Number: 1410564-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR