2-(4-Bromo-2-(trifluoromethoxy)phenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

75859-1G 75859-500MG 75859-5G 75859-25G
77473-116EA 415.09 USD
77473-116 77485-734 77496-322 77505-420
2-(4-Bromo-2-(trifluoromethoxy)phenyl)-1,3-dioxolane ≥97%
2-(4-Bromo-2-(trifluoromethoxy)phenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₈BrF₃O₃
MW: 313.0679 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766187
CAS Number: 2807446-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR