1-Bromo-4-fluoro-2-isobutoxy-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

75763-1G 75763-500MG 75763-25G
77473-112EA 415.09 USD
77473-112 77485-730 77505-416
1-Bromo-4-fluoro-2-isobutoxy-5-methylbenzene ≥97%
1-Bromo-4-fluoro-2-isobutoxy-5-methylbenzene

Precaución: For research use only

Formula: C₁₁H₁₄BrFO
MW: 261.131 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766137
CAS Number: 2901083-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR