2-(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

75673-1G 75673-500MG 75673-5G 75673-25G
77473-108EA 128.75 USD
77473-108 77485-726 77496-314 77505-412
2-(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)propan-2-ol ≥95%
2-(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₀H₉BrF₄O
MW: 301.0754 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766097
CAS Number: 2901082-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR