tert-Butyl 2-(2-Bromopyridin-4-yl)acetate ≥95%

Supplier: AOBChem USA

Warning

75648-1G 75648-500MG 75648-5G
77473-100EA 515 USD
77473-100 77485-718 77496-306
tert-Butyl 2-(2-Bromopyridin-4-yl)acetate ≥95%
tert-Butyl 2-(2-Bromopyridin-4-yl)acetate

Precaución: For research use only

Formula: C₁₁H₁₄BrNO₂
Storage Temperature: Ambient
CAS Number: 1104643-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR