tert-Butyl 2-(6-bromopyridin-2-yl)acetate ≥95%

Supplier: AOBChem USA

Warning

75644-1G 75644-500MG 75644-5G
77473-096EA 482.04 USD
77473-096 77485-714 77496-302
tert-Butyl 2-(6-bromopyridin-2-yl)acetate ≥95%
tert-Butyl 2-(6-bromopyridin-2-yl)acetate

Precaución: For research use only

Formula: C₁₁H₁₄BrNO₂
MW: 272.1383 g/mol
Storage Temperature: Ambient
CAS Number: 1266119-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR