1-(3,5-Dichloropyridin-4-yl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

75604-1G 75604-500MG 75604-5G
77473-092EA 683.92 USD
77473-092 77485-710 77496-298
1-(3,5-Dichloropyridin-4-yl)-2,2-difluoroethanone ≥97%
1-(3,5-Dichloropyridin-4-yl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₇H₃Cl₂F₂NO
Storage Temperature: Ambient
MDL Number: MFCD34815920
CAS Number: 2229281-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR