3-Chloro-4-fluoro-5-isobutoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

75500-1G 75500-500MG 75500-5G 75500-25G
77473-084EA 415.09 USD
77473-084 77485-702 77496-290 77505-398
3-Chloro-4-fluoro-5-isobutoxybenzaldehyde ≥97%
3-Chloro-4-fluoro-5-isobutoxybenzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₂ClFO₂
MW: 230.6631 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766031
CAS Number: 2807443-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR