4-Chloro-2,6-dimethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

75344-1G 75344-500MG 75344-5G 75344-25G
77473-080EA 284.28 USD
77473-080 77485-698 77496-286 77505-394
4-Chloro-2,6-dimethoxybenzaldehyde ≥97%
4-Chloro-2,6-dimethoxybenzaldehyde

Precaución: For research use only

Formula: C₉H₉ClO₃
MW: 200.6189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28128695
CAS Number: 1476848-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR