4-(Difluoromethyl)-3-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

75095-1G 75095-500MG 75095-5G
77473-072EA 806.49 USD
77473-072 77485-690 77496-278
4-(Difluoromethyl)-3-fluorobenzaldehyde ≥97%
4-(Difluoromethyl)-3-fluorobenzaldehyde

Precaución: For research use only

Formula: C₈H₅F₃O
Storage Temperature: Ambient
MDL Number: MFCD28519660
CAS Number: 1780417-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR