2-(2,3-Dichloropyridin-4-yl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

74895-1G 74895-500MG 74895-5G 74895-25G
77473-068EA 272.95 USD
77473-068 77485-686 77496-274 77505-386
2-(2,3-Dichloropyridin-4-yl)propan-2-ol ≥97%
2-(2,3-Dichloropyridin-4-yl)propan-2-ol

Precaución: For research use only

Formula: C₈H₉Cl₂NO
MW: 206.069 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765655
CAS Number: 1305208-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR