2,3-Dichloro-4-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

74893-1G 74893-500MG 74893-5G 74893-25G
77473-064EA 255.44 USD
77473-064 77485-682 77496-270 77505-382
2,3-Dichloro-4-(methylthio)pyridine ≥97%
2,3-Dichloro-4-(methylthio)pyridine

Precaución: For research use only

Formula: C₆H₅Cl₂NS
MW: 194.082 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26127745
CAS Number: 35114-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR