1-Bromo-2,3-dichloro-4-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

74729-1G 74729-500MG 74729-5G 74729-25G
77473-056EA 415.09 USD
77473-056 77485-674 77496-262 77505-376
1-Bromo-2,3-dichloro-4-(methoxymethoxy)benzene ≥97%
1-Bromo-2,3-dichloro-4-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₈H₇BrCl₂O₂
MW: 285.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765589
CAS Number: 2901085-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR