3-Bromo-4-fluoro-5-methoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

74530-1G 74530-500MG 74530-5G
77473-052EA 452.17 USD
77473-052 77485-670 77496-258
3-Bromo-4-fluoro-5-methoxybenzoic acid ≥95%
3-Bromo-4-fluoro-5-methoxybenzoic acid

Precaución: For research use only

Formula: C₈H₆BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD34551501
CAS Number: 1781953-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR