1-(2-Bromo-5-fluoropyridin-4-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

74507-1G 74507-500MG 74507-5G 74507-25G
77473-048EA 167.89 USD
77473-048 77485-666 77496-254 77505-370
1-(2-Bromo-5-fluoropyridin-4-yl)ethanone ≥95%
1-(2-Bromo-5-fluoropyridin-4-yl)ethanone

Precaución: For research use only

Formula: C₇H₅BrFNO
Storage Temperature: Refrigerator
MDL Number: MFCD15526922
CAS Number: 1114523-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR