1-(3-Chloro-4-fluoro-5-(methylthio)phenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

74417-1G 74417-500MG 74417-5G 74417-25G
77473-040EA 428.48 USD
77473-040 77485-658 77496-246 77505-362
1-(3-Chloro-4-fluoro-5-(methylthio)phenyl)ethanol ≥97%
1-(3-Chloro-4-fluoro-5-(methylthio)phenyl)ethanol

Precaución: For research use only

Formula: C₉H₁₀ClFOS
MW: 220.692 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765459
CAS Number: 2807474-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR