(4-Bromo-2,5-dichlorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

74317-1G 74317-500MG 74317-5G 74317-25G
77473-036EA 415.09 USD
77473-036 77485-654 77496-242 77505-358
(4-Bromo-2,5-dichlorophenyl)(methyl)sulfane ≥97%
(4-Bromo-2,5-dichlorophenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₇H₅BrCl₂S
Storage Temperature: Ambient
MDL Number: MFCD34765385
CAS Number: 2168882-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR