tert-Butyl (5-bromo-2-fluorophenyl)carbamate ≥95%

Supplier: AOBChem USA

Warning

74142-1G 74142-500MG 74142-5G 74142-25G
77473-032EA 103 USD
77473-032 77485-650 77496-238 77505-354
tert-Butyl (5-bromo-2-fluorophenyl)carbamate ≥95%
tert-Butyl (5-bromo-2-fluorophenyl)carbamate

Precaución: For research use only

Formula: C₁₁H₁₃BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD21170356
CAS Number: 935285-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR