1-(2-Fluoro-5-isobutoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

74111-1G 74111-500MG 74111-5G 74111-25G
77473-028EA 374.92 USD
77473-028 77485-646 77496-234 77505-350
1-(2-Fluoro-5-isobutoxyphenyl)ethanone ≥97%
1-(2-Fluoro-5-isobutoxyphenyl)ethanone

Precaución: For research use only

Formula: C₁₂H₁₅FO₂
MW: 210.245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765328
CAS Number: 1556503-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR