2-Fluoro-5-isobutoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

74104-1G 74104-500MG 74104-5G 74104-25G
77473-024EA 284.28 USD
77473-024 77485-642 77496-230 77505-346
2-Fluoro-5-isobutoxybenzaldehyde ≥97%
2-Fluoro-5-isobutoxybenzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD34765323
CAS Number: 2901085-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR