2-(4-Bromo-2-fluoro-3-(methylthio)phenyl)-2-methyl-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

73732-1G 73732-500MG 73732-5G 73732-25G
77473-020EA 229.69 USD
77473-020 77485-638 77496-226 77505-344
2-(4-Bromo-2-fluoro-3-(methylthio)phenyl)-2-methyl-1,3-dioxolane ≥97%
2-(4-Bromo-2-fluoro-3-(methylthio)phenyl)-2-methyl-1,3-dioxolane

Precaución: For research use only

Formula: C₁₁H₁₂BrFO₂S
MW: 307.1791 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765127
CAS Number: 2807450-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR