3,5-Dimethoxy-4-(methoxymethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

73585-1G 73585-500MG 73585-5G
77473-012EA 898.16 USD
77473-012 77485-630 77496-218
3,5-Dimethoxy-4-(methoxymethyl)benzaldehyde ≥97%
3,5-Dimethoxy-4-(methoxymethyl)benzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₄O₄
MW: 210.226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765040
CAS Number: 55687-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR