2-Methoxy-3-methylisonicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

73519-1G 73519-500MG 73519-5G
77473-008EA 630.36 USD
77473-008 77485-626 77496-214
2-Methoxy-3-methylisonicotinaldehyde ≥97%
2-Methoxy-3-methylisonicotinaldehyde

Precaución: For research use only

Formula: C₈H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD16611498
CAS Number: 1227603-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR