3-Bromo-4-methyl-5-nitrobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

73241-1G 73241-500MG 73241-5G 73241-25G
77473-000EA 203.94 USD
77473-000 77485-618 77496-206 77505-328
3-Bromo-4-methyl-5-nitrobenzonitrile ≥97%
3-Bromo-4-methyl-5-nitrobenzonitrile

Precaución: For research use only

Formula: C₈H₅BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11007944
CAS Number: 1082041-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR