2,2,2-Trifluoro-1-(3-fluoro-2-methoxy-4-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

73206-1G 73206-500MG 73206-5G 73206-25G
77472-996EA 415.09 USD
77472-996 77485-614 77496-202 77505-326
2,2,2-Trifluoro-1-(3-fluoro-2-methoxy-4-methylphenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(3-fluoro-2-methoxy-4-methylphenyl)ethanone

Precaución: For research use only

Formula: C₁₀H₈F₄O₂
MW: 236.163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764814
CAS Number: 2807465-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR