3-Chloro-2-isopropoxy-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

73164-1G 73164-500MG 73164-5G 73164-25G
77472-992EA 415.09 USD
77472-992 77485-610 77496-198 77505-324
3-Chloro-2-isopropoxy-4-methylbenzaldehyde ≥97%
3-Chloro-2-isopropoxy-4-methylbenzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD34764774
CAS Number: 2807466-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR