2-(P-tolyloxy)pyrimidine-5-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

72969-1G 72969-500MG 72969-5G
77472-980EA 481.01 USD
77472-980 77485-598 77496-186
2-(P-tolyloxy)pyrimidine-5-carbaldehyde ≥95%
2-(P-tolyloxy)pyrimidine-5-carbaldehyde

Precaución: For research use only

Formula: C₁₂H₁₀N₂O₂
MW: 214.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26843568
CAS Number: 1554386-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR