tert-Butyl 2,4-dichloronicotinate ≥95%

Supplier: AOBChem USA

Warning

72935-1G 72935-500MG 72935-5G 72935-25G
77472-976EA 187.46 USD
77472-976 77485-594 77496-182 77505-316
tert-Butyl 2,4-dichloronicotinate ≥95%
tert-Butyl 2,4-dichloronicotinate

Precaución: For research use only

Formula: C₁₀H₁₁Cl₂NO₂
MW: 248.106 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22681907
CAS Number: 1262132-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR